Learn More
Schmidlin™ Whisper-0 Nitrogen Generator
Click to view available options
Flow Rate:
120 mL/min.
40 mL/min.
80 mL/min.
Unit Size:
Each
Description
- Increase efficiency and improve safety and performance
- On-demand supply 24/7
- Flow rate at 8 barg (116 psig)
- Purity: LC/MS grade
- Proprietary membrane technology
- No noise, no moving parts and no electricity
- Ideal for all LC/MS applications
Specifications
Specifications
| Depth (Metric) | 30 cm |
| Flow Rate | 40 mL/min. |
| Height (Metric) | 130 cm |
| Max. Ambient Operating Temperature | 35°C |
| Outlet Pressure | 7 bar |
| Width (English) | 18.9 in. |
| Width (Metric) | 48 cm |
| Connections | Inlet: 3/8 in., Outlet: 1/4 in. |
| Nitrogen Purity | >97% at 40 L/min., >99% at 20 L/min. |