missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ White Polypropylene PE Foam/PTFE Cap

Product Code. 15523271 Shop All Fisherbrand Products
Click to view available options
Neck Finish:
20-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
83-400
89-400
Quantity:
100/Pk.
144/Cs.
288/Pk.
500/Cs.
Unit Size:
Pack of 144
Pack of 288
Pack of 500
This item is not returnable. View return policy

Product Code. 15523271

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Economical alternative to Thermoset and Phenolic PTFE lined caps. Fisherbrand™ White Polypropylene PE Foam/PTFE Cap have a PTFE facing, backed by F217 liners.

Polypropylene (PP) caps with F217 & PTFE liners are an economical alternative to Thermoset & Phenolic PTFE lined caps. They have a PTFE facing, backed by F217 liners. The F217 liners consist of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE). PTFE/F217 liners are very resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference.

  • Features F217 liners which consists of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
  • PTFE/F217 liners are resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference

Specifications

Color White
Description Screw cap
Material Polypropylene
No. per Case 100/Pk.
Packaging Packed in 5 bags of 100
Quantity 100/Pk.
Sterility Sterile
Liner Material PE Foam/PTFE
Neck Finish 33-400
Certifications/Compliance ISO 9001
Shape Round
Product Type PE Foam/PTFE-lined Cap
For Use With (Equipment) Bottle
Closure Material Polypropylene
Closure Type Screw Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.