Learn More
Honeywell™ White Range Super Mont Blanc S2 Shoes
Honeywell™ White Range Super Mont Blanc S2 Shoes are a slip-on ankle boot with a 200J toe cap.
Brand: Honeywell™ 6240748-39/7
Additional Details : Weight : 0.95400kg
Description
- Offers optimal protection
- White treated leather upper with limited stitching
- Resistant to liquid projections and easy to clean
Specifications
European Directive: 89/686/EEC - EC PPE Category II - Standard: EN 345-1 - Marking : S2 Quality Assurance: ISO 9001:2008 | |
Leather | |
White, double density, injected, perforated | |
Metallic 200J Toe Cap |
White | |
39 | |
Polyurethane | |
Safety Footwear |