Learn More
Cole-Parmer™ Wide-Range Pressure Relief Valve
Product Code. 11839642
Click to view available options
Material:
Polypropylene
Unit Size:
Each
Product Code. 11839642
Brand: Cole-Parmer™ 0135066
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Designed for use with Cole-Parmer tubing. These valves are ideal for use with liquid applications.
  • Valves with connections of 1/4 in. or 1/2 in. have a diaphragm made of Viton™ and withstand a maximum inlet pressure of 210psig
  • All other valves have a PTFE shaft design and withstand a maximum inlet pressure of 140psig
  • Shaft design prevents emergency shutdown with a three-seal system that will vent liquid to warn of approaching maintenance
  • All valves are recommended for liquid applications
Specifications
Material Polypropylene
Inlet 1/2 in. NPT(F)
Outlet Port 1/2 in. NPT(F)
Maximum Pressure 210 psig
Product Type Wide-range Pressure Relief Valve