Learn More
Motic™ Widefield Eyepiece for SMZ160
Click to view available options
Magnification Power:
10X
15X
20X
Unit Size:
Each
Description
- Widefield eyepiece for SMZ160 microscope.
- Comes in sizes WF10X/20mm, WF15X/15mm, and WF20X/10mm.
Specifications
Specifications
| Type | Eyepiece |
| Autoclavable | Not Autoclavable |
| Field of View | 10 mm |
| For Use With (Equipment) | SMZ-160 Head |
| Lens Type | Achromatic |
| Magnification Power | 20X |
| Warranty | 5 years |