Learn More
Motic™ Widefield Eyepiece for SMZ160
Product Code. 17887285
Click to view available options
Magnification Power:
10X
15X
20X
Unit Size:
Each
Product Code. 17887285
Brand: Motic™ 1101001403911
Only null left
Add to basket
Add to basket
The Widefield eyepiece is compatible with the SMZ160 microscope.
  • Widefield eyepiece for SMZ160 microscope.
  • Comes in sizes WF10X/20mm, WF15X/15mm, and WF20X/10mm.
Specifications
Type Eyepiece
Autoclavable Not Autoclavable
Field of View 20 mm
For Use With (Application) observation via SMZ-160 stereomicroscope
For Use With (Equipment) SMZ-160
Lens Type Achromatic
Magnification Power 10X
Required Accessories SMZ-160 Head
Sterility Non-sterile
Warranty 5 years