Learn More
Bel-Art™ SP Scienceware™ Wilmad™ Disposable NMR Tube Caps, 10.0mm Dia., Red
Different colors help track samples based on various reference solvents.
Brand: Bel-Art™ 66010-0102
Additional Details : Weight : 0.04310kg
Description
Alert: When using Chloroform or Acetone as a reference solution, please avoid direct contact between the sample and the disposable cap, and avoid long term storage over 2 hours. If these conditions cannot be avoided, please use PTFE tube caps.Specifications
100 per pack | |
Model: Wilmad™ 521-C-100 | |
Red | |
Model: Wilmad™ 521-C-100 |
10.0 mm | |
Polyethylene | |
NMR Spectroscopy |