Learn More
Bel-Art™ SP Scienceware™ Wilmad™ Disposable NMR Tube Caps, 10.0mm Dia., Red
Different colors help track samples based on various reference solvents.
Brand:  Bel-Art™ 66010-1002
Additional Details : Weight : 0.21540kg
 View more versions of this product
Product Code. 15378738
  • 88.90€ / Pack of 1000
Estimated Shipment: 31-07-2024
to see stock.
Add to basket
Description
Description
Alert: When using Chloroform or Acetone as a reference solution, please avoid direct contact between the sample and the disposable cap, and avoid long term storage over 2 hours. If these conditions cannot be avoided, please use PTFE tube caps.
Specifications
Specifications
1000 per pack
Model: Wilmad™ 521-C-1000
Red
Model: Wilmad™ 521-C-1000
10.0 mm
Polyethylene
NMR Spectroscopy