missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Double Layered NMR Tubes for Toxic Samples

Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Double Layered NMR Tubes provides extra protection when working with toxic samples.

Brand:  Bel-Art™ SP Scienceware™ 66000-0002

Additional Details : Weight : 0.04310kg

 View more versions of this product

Product Code. 15378558

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

  • Inner and outer tubes are designed with an approximately 50 μm gap between them
  • Made of Pyrex™ glass
  • Ideal for variable temperature studies
Specifications

Specifications

Outer Tube
4.07 mm
Flat Bottom
5 mm
600 MHz
516-O-5
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions