Learn More
Wilmad-LabGlass™ SP Scienceware™ Rotor and Caps for Bruker MAS Probe
Product Code. 15399028 Shop All SP Scienceware Products
Click to view available options
Model:
520-1A
528-PP-4VAR
For Use With (Equipment):
For Bruker MAS 3.2mm Probe
For Bruker MAS 7mm Probe
Unit Size:
Each
Product Code. 15399028
Brand: Wilmad-LabGlass™ 668090581
Only null left
Add to basket
Add to basket
Provide solution for analysis of solid samples
  • Manufactured from high-purity Zirconia
  • Caps fitted with O-rings for improved sealing
  • MAS Probe and caps compatible for most solid state NMR spectrometers
  • Thoroughly inspected before and after precision-machining process ensures no material irregularities
  • Spin testing performed to highest specified spinning speed, assuring performance without overspinning the rotor
Specifications
For Use With (Equipment) For Bruker MAS 3.2mm Probe
Material Vespel
Description Bottom plug
For Use With (Application) Solid state NMR spectrometers
Model 520-1A