missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Winter Proof Driver Gloves
Honeywell™ Winter Proof Driver Gloves for medium handling in a dry or damp environment.
Brand: Honeywell™ 2001617-08
Additional Details : Weight : 0.09900kg
Description
- Water repellent cowhide grain leather glove
- Brushed fleece lining
- Elasticated at the back
- The leather has been enriched by a special tanning process to give the glove superior water repellent properties
- The lining increases the glove's comfort and cold insulation
- Elasticated at the wrist, the glove offers added support
- The winged thumb makes the glove more anatomical for greater ease of use
- The water repellent treatment enables the glove to be used in damp environments
- Cowhide leather provides good mechanical resistance
- Assembly ensures a longer life while reducing the number of seams
Specifications
0.8 to 1.1 mm | |
Black | |
EN 420-2003 EEC Basic standards, EN 388:2016 Mechanical hazards, EN 511-2006 Cold protection |
8 | |
26.5 cm |