Learn More
Branson Ultrasonics™ Wire Basket For Washer
Product Code. p-8001376
Click to view available options
Height (Metric):
74 mm
Unit Size:
Each
Product Code. 10079240
Brand: Branson Ultrasonics™ 000019129
Only null left
Specifications
Quantity 1 Each
Type Wire Basket
Depth (Metric) 107 mm
For Use With (Equipment) Ultrasonic Washer, 1510/1800 Branson
Height (Metric) 74 mm
Material Stainless Steel
Width (Metric) 125 mm
Model 1510/1800