Learn More
Haleco™ NeutraBase Wood Sodium Phosphate/Cellulose
Product Code. 13205259
Click to view available options
Packaging Quantity:
9.55 kg
Unit Size:
Each
Product Code. 13205259
Brand: Haleco™ JLN100B
Only null left
Add to basket
Add to basket
For neutralizing and solidifying bases for easy and safe cleaning.
  • Makes the spill gel together and decreases the pH
  • Color indicator changes from white to violet during the neutralization, then returns to white once the neutralization has finished
  • Non-toxic
  • Incinerable
  • Ideal for the chemical industry and laboratories
  • Provided in a plastic bucket
Specifications
Color White to violet
Packaging Quantity 9.55 kg
Weight (Metric) 9.55 kg