Learn More
Brady™ WorkHorse™ B-459 Matte Polyester BMP71 Labels
For use in commercial/industrial applications. Brady B-459 matte polyester label with permanent acrylic adhesive are for use with BMP71 printer.
Brand:  Brady™ 114797
Additional Details : Weight : 0.12250kg
 View more versions of this product
Product Code. 11707364
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Specifications
UL Recognized; CSA Accepted; ESD-approved for IEC 61340-5-1 and EN 100015-1
Acrylic, Permanent
Matte
Asset Tracking Label
BMP™71 Printer
500/Roll
1.5 x 0.5 in. (W x H)
-40°C to +100°C
0.079 mm
12.70 x 38.10 mm
12.70 mm
Abrasion, Chemical, Corrosion, Humidity, UV and Weather-Resistant
White
Polyester (B-459)
Asset tracking, Barcoding, Component ID, Equipment ID, General purpose ID, Rating plates
1
Rectangle
38.1 x 12.7 mm (W x H)
0.0031 in.
15.88 mm
38.10 mm