missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Wright Stain
Shop All Epredia Products
Click to view available options
Quantity:
0.95L
Type:
Bottle for Treating Stains
Unit Size:
950mL
Description
- Produces results similar to those of the Giemsa stain with a slightly greater contrast
Specifications
Specifications
| Quantity | 0.95L |
| Type | Bottle for Treating Stains |
Safety and Handling
Product Identifier
- STAIN WRIGHT 950ML
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P321-Specific treatment (see on this label).
- P370+P378-In case of fire: Use dry chemical, CO2, water spray or alcohol-resistant foam to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- MIXTURE LIST-Contains : methanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction