Learn More
Klever X-Change (Narrow Cut Head)
Product Code. p-7241155
Click to view available options
Color:
Blue
Orange
Red
Yellow
Unit Size:
Pack of 12
Product Code. 17748997
Brand: Klever KCJXC20B
Only null left
Add to basket
Add to basket
Recessed blade of the Klever X-Change (Narrow Cut Head) reduces cut injuries and damaged goods.
  • Cuts boxes, film, tape and thousands more applications.
  • Ergonomic 'soft touch' handle reduces hand fatigue and stress.
  • Innovative tape splitter.
  • Adavnced plastic polymers.
  • Highest safety level.
  • Interchangeable, replaceable head.
Specifications
Product Type Klever X Change Narrow Head
For Use With (Application) Cuts Boxes, Film, Tape, and Thousands More Applications
Disposable Reusable
Sterility Non-sterile
Color Blue