missing translation for 'onlineSavingsMsg'
Learn More

uvex X-Craft Pro Low Cut 68002 Black S1 PL PU/TPU Width 11

Product Code. p-200104844 Shop All Uvex Products
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 30245480

Brand: uvex 6800235

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The sporty uvex 1 x-craft safety shoe range takes light applications in trade, industry and logistics to the next level. It is designed for achievers and provides support for a wide variety of activities and working environments.

Total flexibility and support are guaranteed thanks to the sole that features the uvex bionom x principle and an integrated flex zone in the forefoot area. The uvex 1 x-craft also takes abrasion resistance, protection and breathability to the next level thanks to the dynamic scuff cap, technological outer shoe materials and the perforated tongue design. The i-PUREnrj planet midsole ensures consistent energy return a Whether it's in terms of sustainability, comfort, sportiness or innovation, the uvex 1 x-craft is simply the next level of safety shoe.

General features

  • Sporty, especially light S1 PL safety shoe
  • Made from synthetic materials, so suitable for people allergic to chrome
  • All sole materials are free of silicones, plasticisers and other substances that interfere with wetting agents
  • Individually adjustable, elastic laces with quick release

Comfort features

  • Resistant, robust and flexible PUtek outer shoe material in the forefoot area
  • Breathable, light and tear-resistant ripstop material
  • Breathable distance mesh lining
  • Sizes 35 - 40 produced using a woman's last
  • Removable anti-static comfortable insole (article number: 95797-0)

Protection features

  • Shock-absorbing uvex i-PUREnrj planet midsole with energy return, made with 15% recycled granulate from production surpluses
  • Abrasion-resistant uvex x-tended grip planet TPU outsole made with 10% recycled granulate from production surpluses
  • Foamed polyurethane scuff cap
  • Foamed polyurethane heel basket
  • 100% metal-free protective toe cap
  • Flexible textile penetration resistance
  • ESD-certified

Application areas

  • Light applications
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Polycarbonate
Size 35
Sole Material TPU
Toe Material Polymer
Gender Unisex
Resistant To Slipping, Water
Waterproof Water Resistant
Product Type Safety Shoe
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.