Learn More
Laboratoires Anios X12 ALCOOL ISOPROPYLIQUE 70% IP sterile 1L
Brand:  Laboratoires Anios 316.759
X12 ALCOOL ISOPROPYLIQUE 70% IP sterile 1L
Product Code. 16542470
  • ONLINE EXCLUSIVE PRICE
    651.00€
    519.51€ / Pack of 12
    Save 131.49€
    20% Off
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • ANIOS ALCOOL ISOPROPYLIQUE 70% IP STERILE
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : propan-2-ol