missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Xanthosine dihydrate, 99%

Catalog Number p-7077197
26.26 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5968-90-1
C10H12N4O6·2H2O
320.26
AQQAMLPKPXTPOL-GWTDSMLYSA-N
xanthosine dihydrate, unii-1u62i480sp, 9-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purine-2,6-diol dihydrate, xanthosine, dihydrate, xanthosine dihydrate mi, xanthosine,dihydrate, 9-beta-d-ribofuranosyl xanthine dihydrate, 9-, a-d-ribofuranosylxanthine, xanthosine, dihydrate 8ci,9ci
91886582
9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purine-2,6-dione;hydrate
C1=NC2=C(N1C3C(C(C(O3)CO)O)O)NC(=O)NC2=O.O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5968-90-1
C10H12N4O6·2H2O
320.26
AQQAMLPKPXTPOL-GWTDSMLYSA-N
xanthosine dihydrate, unii-1u62i480sp, 9-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purine-2,6-diol dihydrate, xanthosine, dihydrate, xanthosine dihydrate mi, xanthosine,dihydrate, 9-beta-d-ribofuranosyl xanthine dihydrate, 9-, a-d-ribofuranosylxanthine, xanthosine, dihydrate 8ci,9ci
91886582
9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purine-2,6-dione;hydrate
C1=NC2=C(N1C3C(C(C(O3)CO)O)O)NC(=O)NC2=O.O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Merck Index 14,10065
Specific Rotation Condition −53.00 (22.00°C c=8, 0.3N NaOH)
Specific Rotation -53
Formula Weight 320.26
Percent Purity 99%
Chemical Name or Material Xanthosine dihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'