Learn More
Waters Corporation XBridge BEH Amide XP Method Validation Kit
Product Code. 17153612
Encompass
Click to view available options
Length (Metric):
50 mm
100 mm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
Unit Size:
Pack of 3
Product Code. 17153612
Brand: Waters Corporation 186006227
This method validation kit consists of three batches of XBridge BEH Amide columns packed with three different batches of sorbent.
  • Includes three batches of XBridge BEH Amide columns packed with three different batches of sorbent.
Specifications
Diameter (Metric) 2.1 mm
Length (Metric) 50 mm
Max. Pressure 18000 psi (1240 Bar)
Particle Shape Spherical
Particle Size 2.5 μm
Pore Size 130 Å
Quantity 3/Pk.
USP Type L68
Endcapped No
Type eXtended Performance (XP) Method Validation Kit