Learn More
Waters Corporation XBridge Prep Shield RP18 Column
XBridge BEH Shield RP18 columns complement the XBridge BEH C18 and C8 phases and provide a necessary and powerful tool for HPLC method development.
457.50€ - 27900.00€
Specifications
Max. Pressure 6000 psi (415 Bar)
Particle Shape Spherical
Pore Size 130 Å
Quantity 1/Ea.
USP Type L1
View More Specs
Products 22
Product Code Brand Diameter (Metric) Length (Metric) Particle Size Price Quantity & Availability  
Product Code Brand Diameter (Metric) Length (Metric) Particle Size Price Quantity & Availability  
13203270
View Documents Promotion Details
Waters Corporation
186003939
50 mm 100 mm 5 μm
16050.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
13124989
View Documents Promotion Details
Waters Corporation
186003262
30 mm 75 mm 5 μm
7995.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
13263260
View Documents Promotion Details
Waters Corporation
186003935
50 mm 50 mm 5 μm
11745.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
13243300
View Documents Promotion Details
Waters Corporation
186003988
10 mm 10 mm 10 μm
457.50€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
13273300
View Documents Promotion Details
Waters Corporation
186003991
19 mm 10 mm 10 μm
772.50€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
13283300
View Documents Promotion Details
Waters Corporation
186003992
19 mm 50 mm 10 μm
2925.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
13293300
View Documents Promotion Details
Waters Corporation
186003993
19 mm 100 mm 10 μm
4860.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
13203310
View Documents Promotion Details
Waters Corporation
186003994
19 mm 150 mm 10 μm
5760.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
13213310
View Documents Promotion Details
Waters Corporation
186003995
19 mm 250 mm 10 μm
7290.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
13223310
View Documents Promotion Details
Waters Corporation
186003996
30 mm 150 mm 10 μm
13050.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
13233310
View Documents Promotion Details
Waters Corporation
186003997
30 mm 250 mm 10 μm
20400.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
13243310
View Documents Promotion Details
Waters Corporation
186003998
50 mm 50 mm 10 μm
11790.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
13253310
View Documents Promotion Details
Waters Corporation
186003999
50 mm 100 mm 10 μm
16050.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
13252930
View Documents Promotion Details
Waters Corporation
186002985
19 mm 50 mm 5 μm
3045.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
10066604
View Documents Promotion Details
Waters Corporation
186002986
19 mm 100 mm 5 μm
5085.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
13262930
View Documents Promotion Details
Waters Corporation
186002987
19 mm 150 mm 5 μm
6090.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
13272930
View Documents Promotion Details
Waters Corporation
186002988
30 mm 50 mm 5 μm
6270.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
13282930
View Documents Promotion Details
Waters Corporation
186002989
30 mm 100 mm 5 μm
9255.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
13292930
View Documents Promotion Details
Waters Corporation
186002990
30 mm 150 mm 5 μm
11595.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
13263310
View Documents Promotion Details
Waters Corporation
186004001
50 mm 150 mm 10 μm
21900.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
13273310
View Documents Promotion Details
Waters Corporation
186004002
50 mm 250 mm 10 μm
27900.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
13233330
View Documents Promotion Details
Waters Corporation
186004022
19 mm 250 mm 5 μm
7815.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
Description
Description
The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns.
Specifications
Specifications
6000 psi (415 Bar)
130 Å
L1
Column
Spherical
1/Ea.