Learn More
Waters Corporation XBridge Prep Shield RP18 Column
XBridge BEH Shield RP18 columns complement the XBridge BEH C18 and C8 phases and provide a necessary and powerful tool for HPLC method development.
457.50€ - 27900.00€
Specifications
Max. Pressure | 6000 psi (415 Bar) |
---|---|
Particle Shape | Spherical |
Pore Size | 130 Å |
Quantity | 1/Ea. |
USP Type | L1 |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Particle Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Particle Size | Price | Quantity & Availability | |||
13203270
|
Waters Corporation
186003939 |
50 mm | 100 mm | 5 μm |
16050.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
13124989
|
Waters Corporation
186003262 |
30 mm | 75 mm | 5 μm |
7995.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
13263260
|
Waters Corporation
186003935 |
50 mm | 50 mm | 5 μm |
11745.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
13243300
|
Waters Corporation
186003988 |
10 mm | 10 mm | 10 μm |
457.50€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
13273300
|
Waters Corporation
186003991 |
19 mm | 10 mm | 10 μm |
772.50€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
13283300
|
Waters Corporation
186003992 |
19 mm | 50 mm | 10 μm |
2925.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
13293300
|
Waters Corporation
186003993 |
19 mm | 100 mm | 10 μm |
4860.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
13203310
|
Waters Corporation
186003994 |
19 mm | 150 mm | 10 μm |
5760.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
13213310
|
Waters Corporation
186003995 |
19 mm | 250 mm | 10 μm |
7290.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
13223310
|
Waters Corporation
186003996 |
30 mm | 150 mm | 10 μm |
13050.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
13233310
|
Waters Corporation
186003997 |
30 mm | 250 mm | 10 μm |
20400.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
13243310
|
Waters Corporation
186003998 |
50 mm | 50 mm | 10 μm |
11790.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
13253310
|
Waters Corporation
186003999 |
50 mm | 100 mm | 10 μm |
16050.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
13252930
|
Waters Corporation
186002985 |
19 mm | 50 mm | 5 μm |
3045.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
10066604
|
Waters Corporation
186002986 |
19 mm | 100 mm | 5 μm |
5085.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
13262930
|
Waters Corporation
186002987 |
19 mm | 150 mm | 5 μm |
6090.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
13272930
|
Waters Corporation
186002988 |
30 mm | 50 mm | 5 μm |
6270.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
13282930
|
Waters Corporation
186002989 |
30 mm | 100 mm | 5 μm |
9255.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
13292930
|
Waters Corporation
186002990 |
30 mm | 150 mm | 5 μm |
11595.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
13263310
|
Waters Corporation
186004001 |
50 mm | 150 mm | 10 μm |
21900.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
13273310
|
Waters Corporation
186004002 |
50 mm | 250 mm | 10 μm |
27900.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
13233330
|
Waters Corporation
186004022 |
19 mm | 250 mm | 5 μm |
7815.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||
Description
The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns.Specifications
6000 psi (415 Bar) | |
130 Å | |
L1 | |
Column |
Spherical | |
1/Ea. | |
Yes |