Learn More
Waters Corporation XTerra MS C18 Column
XTerra MS C18 columns are general purpose, hybrid-based, reversed-phase C18 columns that are designed to be compatible with mass spectrometry applications.
Brand: Waters Corporation 186000600
Additional Details : Weight : 0.50000kg
Description
- These first generation columns provide superior pH stability compared to a silica-based column
Specifications
4.6 mm | |
6000 psi (415 Bar) | |
2.5 μm | |
1/Ea. | |
Yes |
30 mm | |
Spherical | |
125 Å | |
L1 | |
Column |