Learn More
Chemsplash Xtreme 50 SMS Antistatic Disposable Coverall T5/6 Blue
GREENER_CHOICE
Product Code. 30058421
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
Product Code. 30058421
Brand: Chemsplash 2503.300.004
Only null left
Add to basket
Add to basket
Xtreme SMS 50 Anti-Static Disposable Coverall has an elasticated hood, ankles, a knitted cuff, and a two way zip with an adhesive flap to secure over the zip.
Chemsplash Xtreme SMS 50 Anti-Static Disposable Coverall
  • Material: 50GSM Breathable SMS Fabric
  • Features:
    • Anti-Static Protection
    • Elasticated Hood, Back and Ankles
    • Knitted Cuffs
    • Two-Way Zip With Adhesive Zip Flap
    • Silicone and Latex Free
    • PFAS Free
    • Comes Without the Chest Label
The Chemsplash Xtreme SMS 50 Anti-Static Disposable Coverall is made from breathable SMS fabric chemically treated to offer anti-static protection to EN 1149-5:2008 standards. It also meets CAT III Type 5 and 6. With features such as an elasticated hood, ankles, knitted cuffs, and a two-way zip with an adhesive flap, it provides excellent comfort and robust protection against chemical splash and hazardous particles.
Safety Standards: EN13982-1: TYPE 5, EN13034: TYPE 6, EN1149-5: ANTI-STATIC
Specifications
Product Type Hooded Coveralls
Size Small
Gender Unisex
Material SMS Fabric
Color Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Covered Elastic
Cuff Style Knit
Neck Style Hooded
Hood Type 3-panel
Closure Type Ties
Seam Type Stitched
Sterility Non-sterile
Resistance Anti-static, Particle
Latex Free Yes
No. of Pockets None