Learn More
Chemsplash Xtreme 50 SMS T5/6 Coverall with BlueTaped Seams
GREENER_CHOICE
Product Code. 30058398
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
Product Code. 30058398
Brand: Chemsplash 2590.300.004
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
The Xtreme SMS 50 Coverall with Taped Seams is made from breathable 50gsm SMS fabric.
Chemsplash Xtreme SMS 50 Coverall
  • 50GSM Breathable SMS Fabric
  • Fully Taped Seams
  • Elasticated 3 Pce Hood
  • Elasticated Cuffs & Ankles
  • Two Way Zip With Adhesive Zip Flap
  • Silicone and Latex Free
  • PFAS Free
  • Comes without the chest label
Description: The Chemsplash Xtreme SMS 50 Coverall with Taped Seams is made from breathable 50gsm SMS fabric, compliant to Cat III Type 5 and 6. It offers a robust and strong barrier to dry hazards and is especially suitable for prolonged periods of use due to its breathable properties. All seams are bound.
Specifications
Product Type Hooded Coveralls
Size Small
Gender Unisex
Material SMS Fabric
Color Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type Ties
Seam Type Taped
Sterility Non-sterile
Resistance Particle
Latex Free Yes
No. of Pockets None