missing translation for 'onlineSavingsMsg'
Learn More

Xylazine-d6 Hydrochloride, TRC

Product Code. 30449085
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30449085

Brand: Toronto Research Chemicals TRCX7480021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Xylazine-d6 Hydrochloride
CAS 1795020-86-8
Purity Grade Notes HPLC
Molecular Formula C12 D6 H10 N2 S . H Cl
Quantity 1 mg
Synonym N-[2,6-(Dimethyl-d6)phenyl]-5,6-dihydro-4H-1,3-thiazin-2-amine Hydrochloride, 5,6-Dihydro-2-(2,6-xylidino)-4H-1,3-thiazine-d6 Hydrochloride, 2-[2,6-(Dimethyl-d6)phenylamino]-5,6-dihydro-4H-1,3-thiazine Hydrochloride, BAY-Va 1470-d6, Celactal-d6, Narcosyl-d6, Rompun-d6, Xylapan-d6, Xylasol-d6
InChI Formula InChI=1S/C12H16N2S.ClH/c1-9-5-3-6-10(2)11(9)14-12-13-7-4-8-15-12;/h3,5-6H,4,7-8H2,1-2H3,(H,13,14);1H/i1D3,2D3;
SMILES Cl.[2H]C([2H])([2H])c1cccc(c1NC2=NCCCS2)C([2H])([2H])[2H]
IUPAC Name N-[2,6-bis(trideuteriomethyl)phenyl]-5,6-dihydro-4H-1,3-thiazin-2-amine;hydrochloride
Molecular Weight (g/mol) 262.832
Formula Weight 262.118
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.