Learn More
Melford Yeast, Capsules
Product Code. p-200087096
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
Product Code. 30018942
Brand: Melford Y20026500.0
Each capsule contains 1g Yeast Extract.
Each capsule contains 1g Yeast Extract.
Convenient and easy to use - simply add required number of premeasured capsules according to formulation being prepared without weighing or measuring. Water must be 37°C or warmer to completely dissolve gelatin capsule.
Specifications
Quantity 500 g