missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Yeast Extract Agar (Dehydrated)

Product Code. p-200112230
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11923792

Brand: Thermo Scientific™ CM0019B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Carry out plate count of microorganisms in water with this medium while adhering to ISO 1133:2014.

Carry out plate counts of microorganisms in water with Thermo Scientific™ Oxoid™ Yeast Extract Agar (Dehydrated). Yeast Extract Agar is made to the formula described by Windle Taylor1 for the plate count of microorganisms in water and is tested in accordance with ISO 1133:20142. It is a rich, nutritious medium for the recovery of a wide range of bacteria, yeast, and molds.

The usual method of counting heterotrophic bacteria in water is by the poured plate method with Yeast Extract Agar. Separate counts are made of those aerobic mesophilic organisms which form visible colonies in this medium after 24 hours incubation at 37°C and those which form colonies after three days incubation at 22°C. The two methods give different results2.

The organisms growing under these conditions comprise bacteria, yeasts, and molds.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Yeast Extract Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 21.7L medium

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.