missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Yeast Kex-2 Recombinant Protein, PeproTech®

Product Code. p-7246264
Click to view available options
Quantity:
2 x 500 μg
250 μg
50 μg
500 μg
Unit Size:
1mg
250µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 17884703

Brand: Gibco™ 450451MG

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

450-45-1MG will be provided as 2 x 500 μg (450-45-500UG). Recombinant Yeast Kex-2 is a 60.4 kDa protease consisting of 558 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Proteases (also called Proteolytic Enzymes, Peptidases, or Proteinases) are enzymes that hydrolyze the amide bonds within proteins or peptides. Most proteases act in a specific manner, hydrolyzing bonds at, or adjacent to, specific residues, or a specific sequence of residues contained within the substrate protein or peptide. Proteases play an important role in most diseases and biological processes, including prenatal and postnatal development, reproduction, signal transduction, immune response, various autoimmune and degenerative diseases, and cancer. They are also an important research tool, as they are frequently used in the analysis and production of proteins. Kex-2 cleaves at the carboxyl end of the recognition sequences Arg-Arg/X and Lys-Arg/X.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P13134
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 855483
Molecular Weight (g/mol) 60.4
Name Yeast Kex-2
Quantity 2 x 500 μg
Source Insect cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Symbol KEX2
Specific Reactivity Yeast
Conjugate Unconjugated
Recombinant Recombinant
Sequence LPVPAPPMDS SLLPVKEAED KLSINDPLFE RQWHLVNPSF PGSDINVLDL WYNNITGAGV VAAIVDDGLD YENEDLKDNF CAEGSWDFND NTNLPKPRLS DDYHGTRCAG EIAAKKGNNF CGVGVGYNAK ISGIRILSGD ITTEDEAASL IYGLDVNDIY SCSWGPADDG RHLQGPSDLV KKALVKGVTE GRDSKGAIYV FASGNGGTRG DNCNYDGYTN SIYSITIGAI DHKDLHPPYS EGCSAVMAVT YSSGSGEYIH SSDINGRCSN SHGGTSAAAP LAAGVYTLLL EANPNLTWRD VQYLSILSAV GLEKNADGDW RDSAMGKKYS HRYGFGKIDA HKLIEMSKTW ENVNAQTWFY LPTLYVSQST NSTEETLESV ITISEKSLQD ANFKRIEHVT VTVDIDTEIR GTTTVDLISP AGIISNLGVV RPRDVSSEGF KDWTFMSVAH WGENGVGDWK IKVKTTENGH RIDFHSWRLK LFGESIDSSK TETFVFGNDK EEVEPAATES TVSQYSASST SISISATSTS SISIGVETSA IPQTTTASTD PDSDPNTP
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.