Learn More
Melford Yeast without Amino Acids
Click to view available options
Quantity:
250 g
500 g
1 kg
Unit Size:
1kg
250g
500g
Description
For classifying yeasts based on amino acid and carbohydrates requirements.
For classifying yeasts based on amino acid and carbohydrates requirements. Contains all essential vitamins and inorganic salts necessary for the cultivaiton of yeasts. Excludes histidine methionine, tryptophan and carbon source. Also contains <1 mg/L each of vitamins and trace inorganic salts.
Specifications
Specifications
| Quantity | 500 g |