Learn More
YSI™ DO Cap Membrane Kit
Product Code. p-4110770
Click to view available options
Includes:
Black Cap Membrane, 1.0 mil
Unit Size:
Each
Pack of 6
Product Code. 11938886
Brand: YSI™ 059880
Only null left
Add to basket
Add to basket
DO Cap Membrane Kit for use with probes and electrodes.
  • DO Cap Membrane Kit
  • For use with sensors and probes
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Instrument: 85, 550, and 556 MPS. Probes: 5239, 5905, 5010 and Pro Series Polarographic Sensors
Type DO Cap Membrane Kit
Flow Rate 12 in./sec.
Includes Black Cap Membrane, 1.0 mil
Material PTFE
Probe Type Oxygen/Temperature