Learn More
YSI™ Probe Reconditioning Kit
Click to view available options
Includes:
Sanding Tool and 10 discs
Unit Size:
Each
Description
Includes
Sanding tool and 10 discs
- Reconditions 13-299-43 (Model 5739 Field Oxygen/Temperature Probe), 15-176-50 (Model 5718 Dissolved Oxygen Probe), 13-298-80 (Model 5750 BOD Probe), and 13-298-56, -57, and -62 (Model 55 Handheld Dissolved Oxygen Probes)
- Removes tarnish and silver deposits from sensor's gold electrode
- Maintains spherical contour of probe face
Specifications
Specifications
| For Use With (Equipment) | YSI™ 5718, 5739, 5750, 55 DO Probe |
| Type | Reconditioning Kit |
| Includes | Sanding Tool and 10 discs |
| Probe Type | DO |