Learn More
Emma Safety Footwear Zandvoort Metal and Outdoor Working XD-Fit Shoes
Click to view available options
Size:
40
41
42
43
Unit Size:
1 pair
Description
- Water repellant oiled Nubuck uppers
- Full grain Nappa leather/Split leather lining
- Non-woven antistatic insole with antibacterial inlay sole
- Internal steel toe cap
- Antibacterial inlay sole
- GripForce™ Duo/HRO outsole
Tekniske data
Tekniske data
| Color | Brown |
| Liner Material | Full grain Nappa leather/Split leather |
| Material | Leather |
| Size | 40 |
| Sole Features | Non-woven Antistatic sole |
| Sole Material | Polyurethane midsole |
| Toe Material | Steel |
| Product Type | Metal- and Outdoor Work Shoe |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion