Learn More
ZCCHC11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555739
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: CNCIDLSKSP DPDKSTGTDC RSNLETESSH QSVCTDTSAT SCNCKATEDA SDLNDDDNLP TQELYYVFDK FILTSGK Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 75%.
ZCCHC11 Polyclonal antibody specifically detects ZCCHC11 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
ZCCHC11 | |
Polyclonal | |
Unconjugated | |
TUT4 | |
6030404K05Rik; 9230115F04Rik; Kiaa0191; mKIAA0191; PAP associated domain containing 3; PAPD3; PPAPD3; RP23-406B13.1; terminal uridylyl transferase 4; terminal uridylyltransferase 4; terminal uridylyltransferase 4-like; TUT4; TUTase 4; ZCCHC11; Zinc finger CCHC domain-containing protein 11; zinc finger CCHC-type containing 11; zinc finger, CCHC domain containing 11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23318 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5TAX3 | |
TUT4 | |
Recombinant protein corresponding to Human ZCCHC11 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only