missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Zeatin Riboside
Click to view available options
Quantity:
25 mg
50 mg
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
25mg
50mg
Description
Naturally occurring cytokinin riboside.
Naturally occurring cytokinin riboside. Main effects in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence. Chromatographically purified and recrystallized. Plant cell culture tested. Mixed isomers: trans-isomer 97%, cis-isomer >2%.
Specifications
Specifications
| CAS | 6025-53-2 |
| Quantity | 250 mg |
| Molecular Formula | C15H21N5O5 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction