Learn More
Thermo Scientific™ Zeiss 40X Objective, Plan-Neofluar (0.75 NA)
Product Code. 11803804 Shop All Thermo Scientific Products
Click to view available options
Quantity:
Each
Unit Size:
Each
Product Code. 11803804
Brand: Thermo Scientific™ N010099
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Expand the functionality of your Thermo Scientific™ high-content instrument
Add a wide range of magnifications using Carl Zeiss™ and Olympus™ Objective Lenses for Thermo Scientific™ ArrayScan™ HCA Readers and the Thermo Scientific™ CellInsight™ NXT High Content Screening (HCS) Platform.
TRUSTED_SUSTAINABILITY
Specifications
Type Objective Lens
Product Line Zeiss
Description Zeiss 40X Objective, Plan-Neofluar (0.75 NA)
For Use With (Equipment) ArrayScan HCA Readers
Lens Type Dry
Magnification 40x (0.75NA)
Quantity Each