missing translation for 'onlineSavingsMsg'
Learn More

Newtex™ ZetexPlus™ 200 Series Heat Resistant Gloves

Product Code. p-7230401 Shop All Cole-Parmer Products
Click to view available options
Length (English):
14 in.
Unit Size:
1 pair

Product Code. 15123415

Brand: Newtex™ 8636410

Only null left
Add to basket
Edge
Add to basket

Gloves are constructed from ZetexPlus™ Vermiculite Coated Fiberglass Fabric for protection from temperatures up to 1500°F for extended periods of time.

  • Vermiculite coating evenly dissipates heat across the surface of the glove
  • Coating significantly increases temperature rating and abrasion resistance
  • Premium wool liner
  • Double stitched and reinforced
  • Inorganic fibers will not produce hazardous outgassing

Specifications

Coating Material Vermiculite
Color Tan
Material ZetexPlus Fiberglass Fabric
Size Universal
Temperature (English) Max. 1500°F (Extended), 2000°F (Intermittent)
Temperature (Metric) Max 815°C (Extended), 1095°C (Intermittent)
For Use With (Application) Kiln/Furnace Operations, Heat Processing Facilities, and High Temperature Industrial Environments
Glove Cuff Style Open
Liner Material Wool
Series ZetexPlus™ 200
Length (English) 14 in.
Length (Metric) 35.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.