Learn More
ZVG Zellstoff Vertrieb™ Filter Mask with Exhalation Valve
Use the ZVG Zellstoff Vertrieb™ Filter Mask with Exhalation Valve to filter pulp distribution's fine dust. FFP3 certification indicates protection against harmful, toxic, solid and liquid particles,
Brand: ZVG Zellstoff Vertrieb™ 3095301
12.35 EUR valid until 2024-04-30
Use promo code "21637" to get your promotional price.
Additional Details : Weight : 0.33300kg
Description
- Zetmask FFP3 - Protection against harmful, toxic, solid and liquid particles
- Certifications/Compliance: EN 149: 2002
- With exhalation valve
- Foam-padded nose area guarantees a pressure-free seal fit
Specifications
EN 149: 2002, FFP3 | |
Filter Mask | |
Exhalation valve | |
15/Pk. |
Single-use | |
Yes | |
Yes |