Learn More
ZVG Zellstoff Vertrieb™ Filter Mask with Exhalation Valve
Use the ZVG Zellstoff Vertrieb™ Filter Mask with Exhalation Valve to filter pulp distribution's fine dust. FFP3 certification indicates protection against harmful, toxic, solid and liquid particles,
Brand:  ZVG Zellstoff Vertrieb™ 3095301
12.35 EUR valid until 2024-04-30
Use promo code "21637" to get your promotional price.
Additional Details : Weight : 0.33300kg
Product Code. 10496523
  • 195.00€ / Pack of 15
Estimated Shipment: 18-04-2024
to see stock.
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Zetmask FFP3 - Protection against harmful, toxic, solid and liquid particles
  • Certifications/Compliance: EN 149: 2002
  • With exhalation valve
  • Foam-padded nose area guarantees a pressure-free seal fit
Specifications
Specifications
EN 149: 2002, FFP3
Filter Mask
Exhalation valve
15/Pk.
Single-use