Learn More
ZVG™ zetRoll™ Hand Towel Rolls
Product Code. 17823565
Click to view available options
Dimensions (L x W):
130 m x 21 cm
150 m x 20 cm
153 m x 20.3 cm
Material:
Cellulose
Recycled Paper
Unit Size:
6 rolls
Product Code. 17823565
Brand: ZVG™ 1646306
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
The zetRoll™ Hand Towel Roll provides high absorbency and allows for hygienic hand drying. With its dot-to-dot embossing, it is particularly convenient to use.
The high-quality zetRoll™ disposable paper towel rolls are ideal for gastronomy, schools, care facilities, kitchens, and essential at gas stations for thorough removal of dirt, oil, and fuel residues. They provide a quick and convenient way to dry hands or quickly wipe surfaces or objects in the vicinity. All 2-ply and 3-ply paper towel rolls have point-to-point embossing.
  • White
  • High absorbency
  • Hygienic hand drying
  • With point-to-point embossing
Point-to-point embossing: In a novel manufacturing process, the individual layers of the paper towel are bonded together at multiple points across the entire surface. This creates a unified structure that is highly tear-resistant. This process combines the advantages of a thick wipe (stability) with those of a multi-layered towel (high absorbency).
Specifications
Product Type Towel
Length (English) 13.4 in.
Length (Metric) 153 m
Width (English) 8 in.
Width (Metric) 20.3 cm
Material Cellulose
Color White
Disposable Single-use
Dimensions (L x W) 153 m x 20.3 cm
Ply 2-ply