missing translation for 'onlineSavingsMsg'
Learn More

Zinc 68, 68Zn, plasma standard solution, Specpure™, 68Zn 10μg/mL, Thermo Scientific Chemicals

Catalog Number 15406885
Click to view available options
Quantity:
50 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 50 mL
MDL Number MFCD00145542
UN Number UN3264
Grade Specpure™
Physical Form Liquid
Chemical Name or Material Zinc 68, 68Zn, plasma standard solution, 68Zn 10μg/mL

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'