Learn More
Zinc Chloride, ≥97%, Honeywell™ Fluka™
Free-flowing, ACS reagent, anhydrous,≥97%
Brand: Honeywell Fluka 746355-500G
Additional Details : CAS : 7646-85-7 Poids : 0.50000kg Transport : UN number : 2331 Chem class : 8 Pack group : III
Packaging | Poly bottle |
---|---|
Quantity | 500g |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7646-85-7 | |
136.28 | |
JIAARYAFYJHUJI-UHFFFAOYSA-L | |
5727 | |
dichlorozinc |
Cl2Zn | |
MFCD00011295 | |
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
CHEBI:49976 | |
Cl[Zn]Cl |
Specifications
Zinc chloride | |
Cl2Zn | |
MFCD00011295 | |
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
Cl[Zn]Cl | |
136.28 | |
CHEBI:49976 | |
≥97% | |
Odorless | |
851g/L | |
Solid | |
287°C to 304°C | |
500g |
7646-85-7 | |
ZnCl2 | |
UN2331 | |
JIAARYAFYJHUJI-UHFFFAOYSA-L | |
dichlorozinc | |
5727 | |
136.3g/mol | |
2.93g/cm3 | |
Poly bottle | |
1mmHg (428°C) | |
White | |
5 (at 20°C) |