missing translation for 'onlineSavingsMsg'
Learn More

Zinc Chloride, puriss., Honeywell Fluka™

Catalog Number 15651420
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
25 kg
6 x 1 kg
6 x 2.5 kg
This item is not returnable. View return policy
7646-85-7
Cl2Zn
136.28
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
5727
CHEBI:49976
dichlorozinc
Cl[Zn]Cl
This item is not returnable. View return policy
7646-85-7
Cl2Zn
136.28
MFCD00011295
JIAARYAFYJHUJI-UHFFFAOYSA-L
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
5727
CHEBI:49976
dichlorozinc
Cl[Zn]Cl

Puriss., meets analytical specification of BP, Ph. Eur., USP, 98-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 1 kg
Linear Formula ZnCl2
UN Number 2331
Formula Weight 136.3g/mol
Percent Purity 98 to 100.5%
Grade puriss.
Vapor Pressure 1mmHg (428°C)
Chemical Name or Material Zinc Chloride, Meets analytical specification of Ph. Eur.BP, USP
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'