missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Chloride, puriss., Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
25 kg
6 x 1 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 500 g |
Linear Formula | ZnCl2 |
UN Number | 2331 |
Formula Weight | 136.3g/mol |
Percent Purity | 98 to 100.5% |
Grade | puriss. |
Vapor Pressure | 1mmHg (428°C) |
Chemical Name or Material | Zinc Chloride, Meets analytical specification of Ph. Eur.BP, USP |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'