Learn More
Honeywell Fluka™ Zinc Chloride, puriss. p.a. ACS Reagent, Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥98%
Brand: Honeywell Fluka™ 31650-250G
Additional Details : CAS : 7646-85-7 Poids : 0.25000kg Transport : UN number : 2331 Chem class : 8 Pack group : III
Quantity | 250g |
---|
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7646-85-7 | |
136.28 | |
JIAARYAFYJHUJI-UHFFFAOYSA-L | |
5727 | |
dichlorozinc |
Cl2Zn | |
MFCD00011295 | |
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
CHEBI:49976 | |
Cl[Zn]Cl |
Specifications
Zinc Chloride | |
7646-85-7 | |
ZnCl2 | |
2331 | |
JIAARYAFYJHUJI-UHFFFAOYSA-L | |
dichlorozinc | |
5727 | |
136.3g/mol | |
ACS/puriss. p.a. | |
250g |
Reag. Ph. Eur. reag. ISO | |
Cl2Zn | |
MFCD00011295 | |
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
Cl[Zn]Cl | |
136.28 | |
CHEBI:49976 | |
≥98% | |
1mmHg (428°C) |