missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc diethyldithiocarbamate, Zn 17-19.5%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2 kg
Description
Zinc diethyldithiocarbamate is a chelating ligand. It is used as a common accelerator to latex. A fast curing primary or secondary effective ultra-accelerator for natural and synthetic latex compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 178°C to 183°C |
Color | White |
Density | 1.48 g/mL |
pH | 7.8 |
Flash Point | 204°C (400°F) |
Odor | Odorless |
Linear Formula | Zn[CS2N(CH2CH3)2]2 |
Quantity | 2 kg |
UN Number | UN3077 |
Solubility Information | Insoluble in water. Soluble in benzene carbon disulfide and organic liquids. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'