missing translation for 'onlineSavingsMsg'
Learn More

Zinc diethyldithiocarbamate, Zn 17-19.5%, Thermo Scientific Chemicals

Catalog Number p-7049693
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
14324-55-1
C10H20N2S4Zn
361.904
MFCD00064798
RKQOSDAEEGPRER-UHFFFAOYSA-L
zinc diethyldithiocarbamate, ethyl ziram, zinc bis diethyldithiocarbamate, diethyldithiocarbamic acid zinc salt, zinc n,n-diethylcarbamodithioate, ethyl zimate, unii-icw4708z8g, ditiocarb zinc, zdc, zdec
26633
zinc;N,N-diethylcarbamodithioate
CCN(CC)C(=S)[S-].CCN(CC)C(=S)[S-].[Zn+2]
This item is not returnable. View return policy
14324-55-1
C10H20N2S4Zn
361.904
MFCD00064798
RKQOSDAEEGPRER-UHFFFAOYSA-L
zinc diethyldithiocarbamate, ethyl ziram, zinc bis diethyldithiocarbamate, diethyldithiocarbamic acid zinc salt, zinc n,n-diethylcarbamodithioate, ethyl zimate, unii-icw4708z8g, ditiocarb zinc, zdc, zdec
26633
zinc;N,N-diethylcarbamodithioate
CCN(CC)C(=S)[S-].CCN(CC)C(=S)[S-].[Zn+2]

Chelating agent

Zinc diethyldithiocarbamate is a chelating ligand. It is used as a common accelerator to latex. A fast curing primary or secondary effective ultra-accelerator for natural and synthetic latex compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 178°C to 183°C
Color White
Density 1.48 g/mL
pH 7.8
Flash Point 204°C (400°F)
Odor Odorless
Linear Formula Zn[CS2N(CH2CH3)2]2
Quantity 500 g
UN Number UN3077
Solubility Information Insoluble in water. Soluble in benzene carbon disulfide and organic liquids.
Formula Weight 361.9
Concentration or Composition (by Analyte or Components) Zn 17 to 19.5%
Physical Form Crystalline
Chemical Name or Material Zinc diethyldithiocarbamate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'