missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc, granular, 30 mesh, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Color | Gray |
Melting Point | 420.0°C |
Boiling Point | 907.0°C |
Density | 7.1400g/mL |
Linear Formula | Zn |
Quantity | 100 g |
Fieser | 01,1276; 02,459; 03,334; 04,574; 05,753; 06,672; 07,426; 08,532; 10,459; 11,98; 12,568; 13,298; 14,111; 16,106; 17,406 |
Merck Index | 15, 10326 |
Formula Weight | 65.38 |
Percent Purity | ≥99.7% |
Show More |
Safety and Handling
Product Identifier
- Zinc, granular, 20 mesh
- Danger
- Pyrophoric solid Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- Substance which\, in contact with water\, emits flammable gases Category 1
- H250-Catches fire spontaneously if exposed to air.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P222-Do not allow contact with air.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P370+P378-In case of fire: Use sand to extinguish.
- P273-Avoid release to the environment.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'