missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Oxide, puriss. p.a., ACS Reagent, ≥99% (KT), Honeywell Fluka™
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Melting Point | ∼1975°C |
Color | White |
pH | ∼7 (at 20°C) |
Quantity | 250 g |
Linear Formula | ZnO |
UN Number | UN3077 |
Formula Weight | 81.39g/mol |
Percent Purity | ≥99% (KT) |
Packaging | Poly Bottle |
Appearance | Powder |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'