Learn More
R&L Enterprises™ Zinc Plated Stand Rods
Product Code. 11715386
Click to view available options
Length (Metric):
500 mm
600 mm
750 mm
1000 mm
Diameter (Metric) Rod:
10 mm
12.5 mm
Unit Size:
Each
Pack of 2
Pack of 5
Product Code. 11715386
Brand: R&L Enterprises™ 3108/11
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
An excellent option for the construction of retort stands in the laboratory. R&L Enterprises™ Zinc Plated Rods are manufactured from mild steel.
  • Threaded
  • Available for a range of diameters and lengths
  • Length of rod does not include the thread
Specifications
Material Zinc Plated Mild Steel
Length (Metric) 500 mm
Diameter (Metric) Rod 10 mm
Product Type Rod