missing translation for 'onlineSavingsMsg'
Learn More

Zinc Standard, 1000mg/L in 2% HNO3 (Assurance Grade), SPEX CertiPrep™

Catalog Number p-220621
Click to view available options
Quantity:
30 mL
250 mL
Packaging:
Poly Bottle
Triple-leached LDPE bottle
This item is not returnable. View return policy
7440-66-6, 7697-37-2, 7732-18-5
Zn
65.38
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
CHEBI:30185
zinc
[Zn]
This item is not returnable. View return policy
7440-66-6, 7697-37-2, 7732-18-5
Zn
65.38
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
CHEBI:30185
zinc
[Zn]

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Zn1000μg/mL

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

CAS Min % 1.0, 5.0, 94.0
CAS Max % 1.0, 5.0, 94.0
Grade Plasma
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLZN2-2Y, -2T, -2X    10,000    PLZN2-3Y, -3X
2-5%
Packaging Triple-leached LDPE bottle
Solvent or Matrix 2% HNO3
Quantity 30 mL
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Chemical Name or Material Zinc Standard
compliance-icons
Product Identifier
  • 1000µg/ml zinc
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
Supplemental information
  • MIXTURE LIST-Contains : nitric acid, zinc
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'