missing translation for 'onlineSavingsMsg'
Learn More

Zinc Standard Solution (Spectro ECON), Chem Lab™

Catalog Number 11929152
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
Concentration 1000μg/mL
Quantity 500 mL
compliance-icons
Product Identifier
  • Zinc standard solution 1000
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Nitric acid 69%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'